siRNA Details
Gene Symbol: USP36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321291.1 | NP_001308220.1 | 14 | 1979 | ||
XM_005257542.2 | XP_005257599.1 | 14 | 2003 | |||
XM_005257543.2 | XP_005257600.1 | 14 | 2003 | |||
XM_011525065.1 | XP_011523367.1 | 14 | 2014 | |||
XM_011525066.2 | XP_011523368.1 | 14 | 1876 | |||
XM_011525067.2 | XP_011523369.1 | 14 | 1876 | |||
XM_011525068.1 | XP_011523370.1 | 14 | 1917 | |||
XM_011525069.2 | XP_011523371.1 | 14 | 1876 | |||
XM_011525070.1 | XP_011523372.1 | 15 | 1947 | |||
XM_011525071.2 | XP_011523373.1 | 14 | 1876 | |||
XM_017024900.1 | XP_016880389.1 | 14 | 2003 | |||
XM_017024901.1 | XP_016880390.1 | 12 | 1538 | |||
GenBank mRNA | AB040886.1 | - | 14 | 1896 | ||
AK001671.1 | - | 14 | 2014 | |||
AK022840.1 | - | 8 | 2234 | |||
AK225155.1 | - | 9 | 1323 | |||
AK225426.1 | - | 14 | 1878 | |||
AL833835.1 | - | 1 | 199 | |||
BC016487.1 | - | 14 | 1980 | |||
BC027992.1 | - | 14 | 1917 | |||
BC071582.1 | - | 14 | 1979 | 30345255 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
