siRNA Details
Gene Symbol: UBE2D3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300795.1 | NP_001287724.1 | 8 | 2030 | ||
NM_003340.6 | NP_003331.1 | 8 | 2337 | |||
NM_181886.3 | NP_871615.1 | 8 | 2545 | |||
NM_181887.2 | NP_871616.1 | 8 | 2315 | |||
NM_181888.3 | NP_871617.1 | 8 | 2194 | |||
NM_181889.2 | NP_871618.1 | 8 | 2217 | |||
NM_181890.2 | NP_871619.1 | 8 | 2159 | |||
NM_181891.2 | NP_871620.1 | 8 | 2502 | |||
NM_181892.3 | NP_871621.1 | 8 | 2577 | |||
NM_181893.2 | NP_871622.1 | 7 | 2007 | |||
XM_017008584.1 | XP_016864073.1 | 7 | 2369 | |||
GenBank mRNA | BC037894.2 | - | 8 | 2315 | 5275356 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
