siRNA Details
Gene Symbol: UBE2D3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300795.1 | NP_001287724.1 | 8 | 653 | ||
NM_003340.6 | NP_003331.1 | 8 | 960 | |||
NM_181886.3 | NP_871615.1 | 8 | 1168 | |||
NM_181887.2 | NP_871616.1 | 8 | 938 | |||
NM_181888.3 | NP_871617.1 | 8 | 817 | |||
NM_181889.2 | NP_871618.1 | 8 | 840 | |||
NM_181890.2 | NP_871619.1 | 8 | 782 | |||
NM_181891.2 | NP_871620.1 | 8 | 1125 | |||
NM_181892.3 | NP_871621.1 | 8 | 1200 | |||
NM_181893.2 | NP_871622.1 | 7 | 630 | |||
XM_017008584.1 | XP_016864073.1 | 7 | 992 | |||
GenBank mRNA | AK095822.1 | - | 8 | 1115 | ||
AK127304.1 | - | 6 | 3573 | |||
AL833414.1 | - | 7 | 1743 | |||
BC003395.1 | - | 8 | 640 | 3447673 | ||
BC037894.2 | - | 8 | 937 | 5275356 | ||
BC066917.1 | - | 8 | 671 | 4792462 | ||
BQ428985.1 | - | 8 | 635 | |||
BU570661.1 | - | 7 | 585 | |||
DA550074.1 | - | 8 | 572 | |||
U39318.1 | - | 7 | 520 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
