siRNA Details
Gene Symbol: KMT2E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018682.3 | NP_061152.3 | 3 | 537 | ||
NM_182931.2 | NP_891847.1 | 4 | 685 | |||
XM_005250493.1 | XP_005250550.1 | 3 | 604 | |||
XM_011516400.1 | XP_011514702.1 | 5 | 811 | |||
XM_017012435.1 | XP_016867924.1 | 3 | 596 | |||
GenBank mRNA | AF519459.1 | - | 3 | 352 | ||
AK000940.1 | - | 3 | 352 | |||
AY147037.1 | - | 4 | 685 | |||
AY157990.1 | - | 2 | 151 | |||
BC001296.2 | - | 4 | 647 | |||
BC040004.1 | - | 3 | 555 | |||
BC053906.1 | - | 3 | 593 | |||
BC062583.1 | - | 4 | 657 | |||
BC142987.1 | - | 3 | 567 | |||
JQ809698.1 | - | 2 | 151 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
