siRNA Details
Gene Symbol: KMT2E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018682.3 | NP_061152.3 | 17 | 2813 | ||
NM_182931.2 | NP_891847.1 | 18 | 2961 | |||
XM_005250493.1 | XP_005250550.1 | 17 | 2880 | |||
XM_011516400.1 | XP_011514702.1 | 19 | 3087 | |||
XM_017012435.1 | XP_016867924.1 | 18 | 2961 | |||
GenBank mRNA | AF519459.1 | - | 17 | 2628 | ||
AY147037.1 | - | 18 | 2961 | |||
AY157990.1 | - | 15 | 2187 | |||
AY234382.1 | - | 3 | 558 | |||
JQ809698.1 | - | 16 | 2427 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
