siRNA Details
Gene Symbol: Nr3c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008173.3 | NP_032199.3 | 1 | 1225 | ||
GenBank mRNA | AK037733.1 | - | 1 | 1105 | ||
AK155200.1 | - | 3 | 1307 | |||
AK156323.1 | - | 3 | 1301 | |||
BC008154.1 | - | 2 | 1533 | |||
BC016462.1 | - | 3 | 1308 | |||
BC129912.1 | - | 2 | 1203 | 40111802 | ||
BC129913.1 | - | 2 | 1203 | 40111803 | ||
DQ504162.1 | - | 1 | 1209 | |||
HM236293.1 | - | 2 | 1185 | |||
X04435.1 | - | 2 | 1203 | |||
X13358.1 | - | 2 | 1185 | |||
X13359.1 | - | 2 | 1185 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
