siRNA Details
Gene Symbol: Nr3c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008173.3 | NP_032199.3 | 7 | 2097 | ||
GenBank mRNA | AK037733.1 | - | 7 | 1978 | ||
AK155200.1 | - | 9 | 2179 | |||
AK156323.1 | - | 9 | 2173 | |||
BC129912.1 | - | 8 | 2075 | 40111802 | ||
BC129913.1 | - | 8 | 2075 | 40111803 | ||
DQ504162.1 | - | 7 | 2081 | |||
HM236293.1 | - | 8 | 2057 | |||
X04435.1 | - | 8 | 2075 | |||
X13358.1 | - | 8 | 2057 | |||
X13359.1 | - | 8 | 2060 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
