siRNA Details
Gene Symbol: Hdac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008228.2 | NP_032254.1 | 10 | 1040 | ||
GenBank mRNA | BC092070.1 | - | 10 | 1035 | 4217199 | |
BC092285.1 | - | 10 | 1040 | |||
BC108371.1 | - | 10 | 1041 | 4976514 | ||
U80780.1 | - | 10 | 1037 | |||
X98207.1 | - | 10 | 1040 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
