siRNA Details
Gene Symbol: SRD5A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047.3 | NP_001038.1 | 2 | 670 | ||
NM_001324323.1 | NP_001311252.1 | 3 | 1172 | |||
NR_136739.1 | 2 | 670 | ||||
GenBank mRNA | AF052126.1 | - | 2 | 542 | ||
AF113128.1 | - | 3 | 429 | |||
AK297208.1 | - | 3 | 1067 | |||
AK313070.1 | - | 2 | 552 | |||
BC006373.2 | - | 2 | 523 | |||
BC007033.1 | - | 2 | 552 | 3683274 | ||
BC008673.2 | - | 2 | 526 | 3868483 | ||
BT006834.1 | - | 2 | 415 | |||
M32313.1 | - | 2 | 445 |
Gene Symbol: SRD5A1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028597.1 | 1 | 493 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
