siRNA Details
Gene Symbol: Il12rb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008353.2 | NP_032379.2 | 4 | 468 | ||
GenBank mRNA | AK137577.1 | - | 4 | 469 | ||
AK171937.1 | - | 4 | 445 | |||
BC057212.1 | - | 4 | 451 | 5350447 | ||
U23922.1 | - | 4 | 423 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
