siRNA Details
Gene Symbol: HMOX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002133.2 | NP_002124.1 | 4, 5 | 837 | ||
GenBank mRNA | AK313120.1 | - | 4, 5 | 807 | ||
BC001491.2 | - | 4, 5 | 796 | 3504480 | ||
BT019785.1 | - | 3, 4 | 730 | |||
CR456505.1 | - | 4, 5 | 845 | |||
CR541945.1 | - | 4, 5 | 730 | |||
CR541968.1 | - | 4, 5 | 730 | |||
JF323038.1 | - | 4, 5 | 734 | |||
X06985.1 | - | 4, 5 | 810 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
