siRNA Details
Gene Symbol: Map2k3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008928.4 | NP_032954.1 | 12 | 1151 | ||
GenBank mRNA | AK008141.1 | - | 12 | 1154 | ||
AK011002.1 | - | 12 | 1152 | |||
AK150067.1 | - | 12 | 1152 | |||
AK159379.1 | - | 12 | 1150 | |||
AK166255.1 | - | 12 | 1153 | |||
AK166507.1 | - | 12 | 1152 | |||
AK167209.1 | - | 12 | 1152 | |||
AK167442.1 | - | 12 | 1155 | |||
AK168710.1 | - | 12 | 1134 | |||
AK176968.1 | - | 3 | 121 | |||
BC007467.1 | - | 11 | 1090 | 3497714 | ||
D87115.1 | - | 12 | 1164 | |||
X93150.1 | - | 10 | 886 |

More Information
Other Species Targeted

Gene Ontology Categories:
