siRNA Details
Gene Symbol: Map2k3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008928.4 | NP_032954.1 | 9 | 878 | ||
GenBank mRNA | AK008141.1 | - | 9 | 881 | ||
AK011002.1 | - | 9 | 879 | |||
AK053378.1 | - | 9 | 879 | |||
AK150067.1 | - | 9 | 879 | |||
AK159379.1 | - | 9 | 877 | |||
AK166255.1 | - | 9 | 880 | |||
AK166507.1 | - | 9 | 879 | |||
AK167209.1 | - | 9 | 879 | |||
AK167442.1 | - | 9 | 882 | |||
AK168710.1 | - | 9 | 861 | |||
AK185191.1 | - | 3 | 190 | |||
BC007467.1 | - | 8 | 817 | 3497714 | ||
D87115.1 | - | 9 | 891 | |||
X93150.1 | - | 7 | 613 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
