siRNA Details
Gene Symbol: RIPK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003821.5 | NP_003812.1 | 11 | 1773 | ||
XM_005251092.3 | XP_005251149.1 | 10 | 1583 | |||
XM_011517357.2 | XP_011515659.1 | 9 | 1106 | |||
GenBank mRNA | AF027706.1 | - | 11 | 1683 | ||
AF064824.1 | - | 11 | 1701 | |||
AF078530.1 | - | 11 | 1459 | |||
AK075213.1 | - | 11 | 1677 | |||
AK301448.1 | - | 10 | 1532 | |||
AY358813.1 | - | 10 | 1530 | |||
AY358814.1 | - | 11 | 1710 | |||
AY524045.1 | - | 11 | 1546 | |||
BC004553.2 | - | 11 | 1661 | 4026156 | ||
KU178437.1 | - | 11 | 1459 | |||
KU178438.1 | - | 10 | 1305 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
