siRNA Details
Gene Symbol: Rac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009007.2 | NP_033033.1 | 2 | 278 | ||
GenBank mRNA | AK008435.1 | - | 2 | 279 | ||
AK009017.1 | - | 2 | 279 | |||
AK011072.2 | - | 2 | 279 | |||
AK034601.1 | - | 2 | 279 | |||
AK047969.1 | - | 2 | 281 | |||
AK076023.1 | - | 2 | 320 | |||
AK088825.1 | - | 2 | 290 | |||
AK166383.1 | - | 2 | 285 | |||
AK199407.1 | - | 2 | 90 | |||
BC003828.1 | - | 2 | 279 | 3593957 | ||
BC051053.1 | - | 2 | 278 | 5038182 | ||
CX242948.1 | - | 2 | 315 | |||
JN971018.1 | - | 2 | 82 | |||
X57277.1 | - | 2 | 222 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
