siRNA Details
Gene Symbol: HDAC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003883.3 | NP_003874.2 | 15 | 1325 | ||
XM_011537697.2 | XP_011535999.1 | 14 | 1177 | |||
GenBank mRNA | AF005482.1 | - | 13 | 1341 | ||
AF039703.1 | - | 15 | 1281 | |||
BC000614.1 | - | 15 | 1314 | 3345755 | ||
U66914.1 | - | 15 | 1270 | |||
U75696.1 | - | 14 | 1308 | |||
U75697.1 | - | 15 | 1314 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
