siRNA Details
Gene Symbol: Rac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009007.2 | NP_033033.1 | 3 | 352 | ||
GenBank mRNA | AK008435.1 | - | 3 | 353 | ||
AK009017.1 | - | 3 | 353 | |||
AK011072.2 | - | 3 | 353 | |||
AK034601.1 | - | 3 | 353 | |||
AK047969.1 | - | 3 | 355 | |||
AK076023.1 | - | 3 | 394 | |||
AK088825.1 | - | 3 | 364 | |||
AK166383.1 | - | 3 | 359 | |||
AK199407.1 | - | 3 | 164 | |||
BC003828.1 | - | 3 | 353 | 3593957 | ||
BC051053.1 | - | 3 | 352 | 5038182 | ||
CX242948.1 | - | 3 | 389 | |||
JN971018.1 | - | 3 | 156 | |||
X57277.1 | - | 3 | 296 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
