siRNA Details
Gene Symbol: HDAC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003883.3 | NP_003874.2 | 3 | 319 | ||
GenBank mRNA | AF005482.1 | - | 1 | 335 | ||
AF039703.1 | - | 3 | 275 | |||
BC000614.1 | - | 3 | 308 | 3345755 | ||
BM461585.1 | - | 3 | 309 | |||
DB456387.1 | - | 3 | 333 | |||
U66914.1 | - | 3 | 264 | |||
U75696.1 | - | 2 | 302 | |||
U75697.1 | - | 3 | 308 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
