siRNA Details
Gene Symbol: Hdac3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010411.2 | NP_034541.2 | 14 | 1240 | ||
GenBank mRNA | AF074881.1 | - | 14 | 1187 | ||
AF074882.1 | - | 8 | 614 | |||
AF098295.1 | - | 14 | 1165 | |||
AF125536.1 | - | 14 | 1172 | |||
AK037478.1 | - | 1232 | ||||
AK145158.1 | - | 14 | 1206 | |||
BC139300.1 | - | 14 | 1268 | 8862322 | ||
BC139301.1 | - | 14 | 1268 | 8862323 | ||
JN651901.1 | - | 8 | 888 | |||
JN651902.1 | - | 7 | 846 | |||
JN651903.1 | - | 8 | 597 | |||
JN651904.1 | - | 5 | 442 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
