siRNA Details
Gene Symbol: RPS6KA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322227.1 | NP_001309156.1 | 7, 8 | 1065 | ||
NM_001322228.1 | NP_001309157.1 | 5, 6 | 817 | |||
NM_001322229.1 | NP_001309158.1 | 6, 7 | 925 | |||
NM_001322230.1 | NP_001309159.1 | 6, 7 | 925 | |||
NM_001322232.1 | NP_001309161.1 | 5, 6 | 706 | |||
NM_001322233.1 | NP_001309162.1 | 6, 7 | 925 | |||
NM_001322234.1 | NP_001309163.1 | 6, 7 | 962 | |||
NM_001322235.1 | NP_001309164.1 | 7, 8 | 1065 | |||
NM_001322237.1 | NP_001309166.1 | 6, 7 | 724 | |||
NM_001322238.1 | NP_001309167.1 | 4, 5 | 433 | |||
NM_004755.3 | NP_004746.2 | 6, 7 | 925 | |||
NM_182398.2 | NP_872198.1 | 6, 7 | 925 | |||
XM_017021786.1 | XP_016877275.1 | 7, 8 | 964 | |||
GenBank mRNA | AF074393.1 | - | 6, 7 | 875 | ||
AF080000.1 | - | 6, 7 | 891 | |||
AF090421.1 | - | 6, 7 | 874 | |||
AK295266.1 | - | 6, 7 | 724 | |||
AK309925.1 | - | 6, 7 | 949 | |||
BC017187.2 | - | 6, 7 | 876 | 3139114 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
