siRNA Details
Gene Symbol: HDAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004964.2 | NP_004955.2 | 10 | 1127 | ||
XM_011541309.2 | XP_011539611.1 | 10 | 1512 | |||
GenBank mRNA | AB451430.1 | - | 10 | 1064 | ||
AK299241.1 | - | 10 | 1061 | |||
AK313061.1 | - | 10 | 1129 | |||
AY627042.1 | - | 10 | 1071 | |||
BC000301.2 | - | 10 | 1098 | 2820260 | ||
BX648055.1 | - | 3 | 1431 | |||
CR541717.1 | - | 10 | 1064 | |||
D50405.1 | - | 10 | 1127 | |||
U50079.1 | - | 10 | 1174 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
