siRNA Details
Gene Symbol: Il17a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010552.3 | NP_034682.1 | 3 | 389 | ||
GenBank mRNA | AK040420.1 | - | 3 | 390 | ||
BC119303.1 | - | 3 | 389 | 8734052 | ||
BC119309.1 | - | 3 | 389 | 8734058 | ||
U43088.1 | - | 3 | 376 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
