siRNA Details
Gene Symbol: HDAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004964.2 | NP_004955.2 | 3 | 265 | ||
XM_011541309.2 | XP_011539611.1 | 3 | 269 | |||
GenBank mRNA | AB451297.1 | - | 3 | 202 | ||
AB451430.1 | - | 3 | 202 | |||
AK225555.1 | - | 3 | 270 | |||
AK296308.1 | - | 2 | 155 | |||
AK299241.1 | - | 3 | 199 | |||
AK299792.1 | - | 3 | 225 | |||
AK313061.1 | - | 3 | 268 | |||
AY627042.1 | - | 3 | 209 | |||
BC000301.2 | - | 3 | 236 | 2820260 | ||
CR541717.1 | - | 3 | 202 | |||
D50405.1 | - | 3 | 265 | |||
U50079.1 | - | 3 | 312 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
