siRNA Details
Gene Symbol: Jak3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190830.1 | NP_001177759.1 | 18 | 2660 | ||
NM_010589.6 | NP_034719.2 | 18 | 2660 | |||
GenBank mRNA | AK038268.1 | - | 17 | 2471 | ||
AK088365.1 | - | 18 | 2509 | |||
AK142178.1 | - | 18 | 2498 | |||
AK156646.1 | - | 18 | 2482 | |||
AK164520.1 | - | 18 | 2500 | |||
BC105577.1 | - | 17 | 2463 | 3968386 | ||
BC131646.1 | - | 17 | 2471 | 40110146 | ||
BC131647.1 | - | 17 | 2471 | 40110148 | ||
L32955.1 | - | 17 | 2462 | |||
L33768.1 | - | 15 | 3098 | |||
L40172.1 | - | 18 | 2521 |

More Information
Other Species Targeted

Gene Ontology Categories:
