siRNA Details
Gene Symbol: Jak3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190830.1 | NP_001177759.1 | 4 | 614 | ||
NM_010589.6 | NP_034719.2 | 4 | 614 | |||
GenBank mRNA | AK038268.1 | - | 3 | 425 | ||
AK088365.1 | - | 4 | 466 | |||
AK142178.1 | - | 4 | 452 | |||
AK156646.1 | - | 4 | 436 | |||
AK164520.1 | - | 4 | 454 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
