siRNA Details
Gene Symbol: Jak3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190830.1 | NP_001177759.1 | 5, 6 | 778 | ||
NM_010589.6 | NP_034719.2 | 5, 6 | 778 | |||
GenBank mRNA | AK038268.1 | - | 4, 5 | 589 | ||
AK088365.1 | - | 5, 6 | 630 | |||
AK142178.1 | - | 5, 6 | 616 | |||
AK156646.1 | - | 5, 6 | 600 | |||
AK164520.1 | - | 5, 6 | 618 | |||
BC105577.1 | - | 4, 5 | 581 | 3968386 | ||
BC131646.1 | - | 4, 5 | 589 | 40110146 | ||
BC131647.1 | - | 4, 5 | 589 | 40110148 | ||
L32955.1 | - | 4, 5 | 580 | |||
L33768.1 | - | 4, 5 | 574 | |||
L40172.1 | - | 5, 6 | 639 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
