siRNA Details
Gene Symbol: Fut7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177366.1 | NP_001170837.1 | 2 | 1136 | ||
NM_001177367.1 | NP_001170838.1 | 3 | 1221 | |||
NM_001289453.1 | NP_001276382.1 | 4 | 1264 | |||
NM_001289454.1 | NP_001276383.1 | 4 | 1236 | |||
NM_001289455.1 | NP_001276384.1 | 3 | 1159 | |||
NM_001289456.1 | NP_001276385.1 | 4 | 1128 | |||
NM_013524.3 | NP_038552.1 | 3 | 1342 | |||
GenBank mRNA | AK154486.1 | - | 4 | 1243 | ||
BC104732.1 | - | 3 | 1123 | 40046865 | ||
BC105670.1 | - | 2 | 1100 | 40046875 | ||
BC105671.1 | - | 3 | 1185 | 40046874 | ||
BC105672.1 | - | 3 | 1185 | 40046873 | ||
GQ424189.1 | - | 4 | 1236 | |||
GQ424190.1 | - | 4 | 1263 | |||
GQ424191.1 | - | 4 | 1117 | |||
GQ424192.1 | - | 2 | 1451 | |||
GQ424193.1 | - | 3 | 1035 | |||
GQ424194.1 | - | 2 | 1076 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
