siRNA Details
Gene Symbol: Irak4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029926.5 | NP_084202.2 | 7 | 946 | ||
GenBank mRNA | AF445803.1 | - | 7 | 934 | ||
AK028837.1 | - | 7 | 887 | |||
AK029028.1 | - | 7 | 947 | |||
AK139992.1 | - | 935 | ||||
BC051676.1 | - | 7 | 939 | 30017484 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
