siRNA Details
Gene Symbol: Il23a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031252.2 | NP_112542.1 | 4 | 569 | ||
GenBank mRNA | AF301619.1 | - | 4 | 569 | ||
AK043894.1 | - | 1456 | ||||
BC019953.1 | - | 4 | 553 | 3708687 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
