siRNA Details
Gene Symbol: MAP2K3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316332.1 | NP_001303261.1 | 7 | 821 | ||
NM_002756.4 | NP_002747.2 | 6 | 438 | |||
NM_145109.2 | NP_659731.1 | 6 | 696 | |||
XM_005256723.2 | XP_005256780.1 | 7 | 736 | |||
XM_011523958.2 | XP_011522260.1 | 6 | 495 | |||
XM_011523959.1 | XP_011522261.1 | 8 | 703 | |||
XM_017024857.1 | XP_016880346.1 | 6 | 565 | |||
XM_017024858.1 | XP_016880347.1 | 7 | 553 | |||
XM_017024859.1 | XP_016880348.1 | 8 | 893 | |||
GenBank mRNA | AB451226.1 | - | 6 | 431 | ||
AB451349.1 | - | 6 | 431 | |||
AK093838.1 | - | 7 | 740 | |||
AK223490.1 | - | 6 | 571 | |||
AK315505.1 | - | 6 | 641 | |||
BC032478.1 | - | 6 | 631 | 5215093 | ||
CR536514.1 | - | 6 | 431 | |||
CR541793.1 | - | 6 | 431 | |||
D87116.1 | - | 6 | 488 | |||
DA987047.1 | - | 6 | 439 | |||
GQ225578.1 | - | 6 | 431 | |||
L36719.1 | - | 8 | 681 | |||
U66839.1 | - | 6 | 557 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
