siRNA Details
Gene Symbol: Ripk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138952.3 | NP_620402.1 | 2, 3 | 425 | ||
GenBank mRNA | AF461040.1 | - | 2, 3 | 323 | ||
AF487539.1 | - | 2, 3 | 323 | |||
AK159593.1 | - | 2, 3 | 449 | |||
AK172319.1 | - | 2, 3 | 447 | |||
AK172417.1 | - | 2, 3 | 447 | |||
BC050845.1 | - | 2, 3 | 438 | |||
BC058088.1 | - | 2, 3 | 570 | |||
BC069878.1 | - | 2, 3 | 450 | 5316066 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
