siRNA Details
Gene Symbol: MAP2K3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316332.1 | NP_001303261.1 | 10 | 1102 | ||
NM_002756.4 | NP_002747.2 | 9 | 719 | |||
NM_145109.2 | NP_659731.1 | 9 | 977 | |||
XM_005256723.2 | XP_005256780.1 | 10 | 1017 | |||
XM_011523958.2 | XP_011522260.1 | 9 | 776 | |||
XM_011523959.1 | XP_011522261.1 | 11 | 984 | |||
XM_017024857.1 | XP_016880346.1 | 9 | 846 | |||
XM_017024858.1 | XP_016880347.1 | 10 | 834 | |||
XM_017024859.1 | XP_016880348.1 | 11 | 1174 | |||
GenBank mRNA | AB451226.1 | - | 9 | 712 | ||
AB451349.1 | - | 9 | 712 | |||
AK093838.1 | - | 10 | 1021 | |||
AK223490.1 | - | 9 | 852 | |||
AK315505.1 | - | 9 | 922 | |||
BC032478.1 | - | 9 | 912 | 5215093 | ||
CR536514.1 | - | 9 | 712 | |||
CR541793.1 | - | 9 | 712 | |||
D87116.1 | - | 9 | 769 | |||
GQ225578.1 | - | 9 | 712 | |||
L36719.1 | - | 11 | 962 | |||
U66839.1 | - | 9 | 838 |
Gene Symbol: LOC100996792 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011524134.2 | XP_011522436.2 | 2 | 83 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
