siRNA Details
Gene Symbol: Map3k7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009316.1 | NP_033342.1 | 8 | 912 | ||
NM_172688.3 | NP_766276.1 | 8 | 912 | |||
GenBank mRNA | AK046565.1 | - | 8 | 878 | ||
AK053911.1 | - | 8 | 898 | |||
AK133642.1 | - | 2 | 39 | |||
AK137046.1 | - | 8 | 913 | |||
AK159277.1 | - | 8 | 876 | |||
BC006665.1 | - | 8 | 903 | 3499247 | ||
D76446.1 | - | 8 | 912 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
