siRNA Details
Gene Symbol: Map3k7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009316.1 | NP_033342.1 | 17 | 1922 | ||
NM_172688.3 | NP_766276.1 | 16 | 1841 | |||
GenBank mRNA | AK046565.1 | - | 16 | 1807 | ||
AK053911.1 | - | 16 | 1827 | |||
AK133642.1 | - | 10 | 968 | |||
AK137046.1 | - | 16 | 1842 | |||
AK159277.1 | - | 16 | 1805 | |||
BC006665.1 | - | 17 | 1913 | 3499247 | ||
D76446.1 | - | 16 | 1841 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001107920, XM_003750753, XM_006237966, XM_006245614.
Gene Ontology Categories:
