siRNA Details
Gene Symbol: Srd5a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175283.3 | NP_780492.2 | 2 | 652 | ||
GenBank mRNA | AK082819.1 | - | 2 | 544 | ||
BC079863.1 | - | 2 | 630 | 6807475 | ||
BC094503.1 | - | 631 | 5053446 | |||
BC115468.1 | - | 2 | 509 | 40100423 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
