siRNA Details
Gene Symbol: SRD5A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047.3 | NP_001038.1 | 4, 5 | 960 | ||
NM_001324322.1 | NP_001311251.1 | 3, 4 | 793 | |||
NM_001324323.1 | NP_001311252.1 | 5, 6 | 1462 | |||
NR_136739.1 | 5, 6 | 1150 | ||||
GenBank mRNA | AF052126.1 | - | 4, 5 | 832 | ||
AF113128.1 | - | 5, 6 | 719 | |||
AK297208.1 | - | 5, 6 | 1357 | |||
AK297511.1 | - | 3, 4 | 688 | |||
AK313070.1 | - | 4, 5 | 842 | |||
AK315996.1 | - | 3, 4 | 647 | |||
BC006373.2 | - | 4, 5 | 813 | |||
BC007033.1 | - | 4, 5 | 842 | 3683274 | ||
BC008673.2 | - | 4, 5 | 816 | 3868483 | ||
BT006834.1 | - | 4, 5 | 705 | |||
M32313.1 | - | 4, 5 | 735 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
