siRNA Details
Gene Symbol: Prkcq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008859.2 | NP_032885.1 | 8 | 866 | ||
GenBank mRNA | AK037664.1 | - | 8 | 938 | ||
AK085546.1 | - | 8 | 1136 | |||
BC138552.1 | - | 7 | 797 | 8861572 | ||
BC145769.1 | - | 7 | 797 | 40131072 | ||
D11091.1 | - | 8 | 866 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
