siRNA Details
Gene Symbol: HMOX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002133.2 | NP_002124.1 | 5 | 1371 | ||
GenBank mRNA | BC001491.2 | - | 5 | 1330 | 3504480 | |
M23042.1 | - | 1 | 298 | |||
X06985.1 | - | 5 | 1344 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
