siRNA Details
Gene Symbol: Prkcq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008859.2 | NP_032885.1 | 12 | 1324 | ||
GenBank mRNA | AB062122.1 | - | 5 | 722 | ||
AK037664.1 | - | 12 | 1396 | |||
AK085546.1 | - | 12 | 1594 | |||
BC138552.1 | - | 11 | 1255 | 8861572 | ||
BC145769.1 | - | 11 | 1255 | 40131072 | ||
D11091.1 | - | 12 | 1324 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
