siRNA Details
Gene Symbol: FUT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004479.3 | NP_004470.1 | 2 | 2301 | ||
GenBank mRNA | BC086312.1 | - | 2 | 2301 | 6651145 | |
X78031.1 | - | 2 | 1574 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
