siRNA Details
Gene Symbol: Il12a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF128210.1 | - | 3 | 259 | ||
AF128211.1 | - | 3 | 259 | |||
AF128212.1 | - | 3 | 259 | |||
AF128213.1 | - | 3 | 259 | |||
AK153729.1 | - | 1 | 226 | |||
BC116851.1 | - | 3 | 263 | |||
BC116855.1 | - | 3 | 263 | |||
M86672.1 | - | 3 | 361 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
