siRNA Details
Gene Symbol: Hdac3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010411.2 | NP_034541.2 | 14 | 1200 | ||
GenBank mRNA | AF074881.1 | - | 14 | 1147 | ||
AF074882.1 | - | 8 | 574 | |||
AF098295.1 | - | 14 | 1125 | |||
AF125536.1 | - | 14 | 1132 | |||
AK037478.1 | - | 1192 | ||||
AK145158.1 | - | 14 | 1166 | |||
BC139300.1 | - | 14 | 1228 | 8862322 | ||
BC139301.1 | - | 14 | 1228 | 8862323 | ||
JN651901.1 | - | 8 | 848 | |||
JN651902.1 | - | 7 | 806 | |||
JN651903.1 | - | 8 | 557 | |||
JN651904.1 | - | 5 | 402 |

More Information
Other Species Targeted

Gene Ontology Categories:
