siRNA Details
Gene Symbol: Srd5a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK082819.1 | - | 4 | 838 | ||
BC115468.1 | - | 4 | 936 | 40100423 | ||
BC115469.1 | - | 4 | 936 | 40100424 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
