siRNA Details
Gene Symbol: RAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006908.4 | NP_008839.2 | 6 | 2226 | ||
NM_018890.3 | NP_061485.1 | 7 | 2283 | |||
GenBank mRNA | AK054993.1 | - | 6 | 2201 | ||
BC050687.1 | - | 6 | 2173 | 6149377 | ||
D25274.1 | - | 1 | 1117 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
