siRNA Details
Gene Symbol: BCL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000633.2 | NP_000624.2 | 2 | 1031 | ||
NM_000657.2 | NP_000648.2 | 2 | 1031 | |||
XM_011526135.2 | XP_011524437.1 | 1 | 1650 | |||
XM_017025917.1 | XP_016881406.1 | 2 | 1429 | |||
GenBank mRNA | BC027258.1 | - | 2 | 927 | 4511027 | |
EU287875.1 | - | 1 | 538 | |||
M13994.1 | - | 1 | 1996 | |||
M13995.1 | - | 1 | 684 | |||
M14745.1 | - | 1 | 569 | |||
X06487.1 | - | 2 | 1424 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_009741, NM_177410, BC068988, AK049473, BC095964, and others.
Gene Ontology Categories:
