siRNA Details
Gene Symbol: UGT1A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019077.2 | NP_061950.2 | 1 | 420 | ||
GenBank mRNA | AY435142.1 | - | 1 | 420 | ||
DQ383513.1 | - | 1 | 132 | |||
U89507.1 | - | 1 | 420 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
