siRNA Details
Gene Symbol: GGT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005261890.4 | XP_005261947.1 | 8 | 1337 | ||
XM_006724392.3 | XP_006724455.1 | 12 | 2045 | |||
XM_006724395.3 | XP_006724458.1 | 9 | 1497 | |||
XM_006724396.2 | XP_006724459.1 | 5 | 875 | |||
XM_017029179.1 | XP_016884668.1 | 13 | 2726 | |||
XM_017029180.1 | XP_016884669.1 | 12 | 2184 | |||
XM_017029181.1 | XP_016884670.1 | 12 | 2622 | |||
XM_017029183.1 | XP_016884672.1 | 8 | 1313 | |||
XM_017029184.1 | XP_016884673.1 | 5 | 885 | |||
XM_017029185.1 | XP_016884674.1 | 15 | 3507 | |||
XM_017029186.1 | XP_016884675.1 | 18 | 3108 |
Gene Symbol: GGTLC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178311.2 | NP_842563.1 | 6 | 876 | ||
NM_178312.2 | NP_842564.1 | 6 | 804 | |||
XM_005260863.4 | XP_005260920.1 | 5 | 1312 | |||
XM_005260864.4 | XP_005260921.1 | 6 | 1465 | |||
XM_005260865.4 | XP_005260922.1 | 7 | 1384 | |||
XM_011529391.2 | XP_011527693.1 | 5 | 879 | |||
XM_011529392.2 | XP_011527694.1 | 7 | 1323 | |||
XM_011529393.2 | XP_011527695.1 | 6 | 1170 | |||
XM_017028126.1 | XP_016883615.1 | 6 | 793 | |||
GenBank mRNA | BC040904.1 | - | 6 | 876 | 5744021 | |
L20491.1 | - | 6 | 804 | |||
L20492.1 | - | 5 | 854 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
