siRNA Details
Gene Symbol: PI4KAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003700.1 | 17 | 2168 | |||
GenBank mRNA | AK298744.1 | - | 17 | 2310 | ||
BC020225.1 | - | 17 | 2167 |
Gene Symbol: PI4KAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK002141.1 | - | 3 | 2321 | ||
AK024034.1 | - | 4 | 1694 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
