siRNA Details
Gene Symbol: AMY2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020978.4 | NP_066188.1 | 5 | 1060 | ||
GenBank mRNA | AK095605.1 | - | 4 | 1730 | ||
AK127047.1 | - | 4 | 2499 | |||
BC011179.1 | - | 5 | 752 | 4150646 | ||
BC020861.1 | - | 4 | 524 | 4551745 | ||
M24895.1 | - | 3 | 434 |
Gene Symbol: AMY1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008221.1 | NP_001008222.1 | 4 | 602 | ||
NM_004038.3 | NP_004029.2 | 4 | 683 | |||
GenBank mRNA | AK292341.1 | - | 4 | 601 | ||
AK313275.1 | - | 4 | 603 | |||
BC063129.1 | - | 4 | 612 | 30327425 | ||
BC069347.1 | - | 3 | 388 | 7262297 | ||
BC069463.1 | - | 3 | 388 | 7262309 | ||
BC070302.1 | - | 4 | 608 | |||
BC092444.1 | - | 4 | 579 | 30349929 | ||
BC132985.1 | - | 4 | 558 | 40147007 | ||
BC132987.1 | - | 4 | 558 | 40147009 | ||
BC132995.1 | - | 4 | 557 | 40147017 | ||
BC132997.1 | - | 4 | 558 | 40147019 | ||
BC144452.1 | - | 4 | 558 | 9052978 |
Gene Symbol: AMY1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008218.1 | NP_001008219.1 | 4 | 660 | ||
XM_011541262.1 | XP_011539564.1 | 4 | 603 | |||
GenBank mRNA | CB957894.1 | - | 2 | 201 |
Gene Symbol: AMY1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008219.1 | NP_001008220.1 | 4 | 682 | ||
XM_011541263.1 | XP_011539565.1 | 4 | 603 | |||
XM_017001058.1 | XP_016856547.1 | 4 | 520 | |||
GenBank mRNA | CB955582.1 | - | 4 | 471 | ||
CD690933.1 | - | 5 | 613 |
Gene Symbol: AMY2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000699.3 | NP_000690.1 | 3 | 497 | ||
GenBank mRNA | AK223463.1 | - | 3 | 412 | ||
BC007060.1 | - | 3 | 452 | 3949799 | ||
BC146997.1 | - | 3 | 497 | 9056925 | ||
M28443.1 | - | 3 | 401 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
