siRNA Details
Gene Symbol: AMY2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020978.4 | NP_066188.1 | 6 | 1302 | ||
GenBank mRNA | AK095605.1 | - | 5 | 1972 | ||
AK127047.1 | - | 5 | 2741 | |||
BC011179.1 | - | 6 | 994 | 4150646 | ||
BC020861.1 | - | 5 | 766 | 4551745 | ||
M24895.1 | - | 4 | 676 |
Gene Symbol: AMY2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000699.3 | NP_000690.1 | 4 | 739 | ||
GenBank mRNA | AK223463.1 | - | 4 | 654 | ||
BC007060.1 | - | 4 | 694 | 3949799 | ||
BC146997.1 | - | 4 | 739 | 9056925 | ||
M28443.1 | - | 4 | 643 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
